Skip to Content
Merck
All Photos(1)

Key Documents

673013

Sigma-Aldrich

2-Benzyloxyphenylmagnesium bromide solution

1.0 M in THF

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$471.00

$471.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 ML
$471.00

About This Item

Empirical Formula (Hill Notation):
C13H11BrMgO
CAS Number:
Molecular Weight:
287.43
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$471.00


Check Cart for Availability

Request a Bulk Order

reaction suitability

reaction type: Grignard Reaction

concentration

1.0 M in THF

density

1.047 g/mL at 25 °C

functional group

phenyl

SMILES string

Br[Mg]c1ccccc1OCc2ccccc2

InChI

1S/C13H11O.BrH.Mg/c1-3-7-12(8-4-1)11-14-13-9-5-2-6-10-13;;/h1-9H,11H2;1H;/q;;+1/p-1

InChI key

VELKJPFZDYXBDE-UHFFFAOYSA-M

Application

2-Benzyloxyphenylmagnesium bromide can be used:
  • As a substrate in the iron-catalyzed coupling reaction of aryl Grignard reagents with unprotected bromophenols.[1]
  • As a substrate in the cross-coupling of Grignard reagents using diaryl titanates and ferric chloride.[2]
  • In one of the key synthetic steps for the total synthesis of an antibiotic caboxamycin.[3]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-4.0 - 5.0 °F - closed cup

Flash Point(C)

-20 - -15 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iron catalyzed oxidative assembly of N-heteroaryl and aryl metal reagents using oxygen as an oxidant
Liu KM, et al.
Chemical Communications (Cambridge, England), 51(6), 1124-1127 (2015)
Iron-Catalyzed Room Temperature Cross-Couplings of Bromophenols with Aryl Grignard Reagents
Xu L, et al.
Advanced Synthesis & Catalysis, 361(23), 5421-5427 (2019)
Iron-catalyzed oxidative biaryl cross-couplings via mixed diaryl titanates: significant influence of the order of combining aryl Grignard reagents with titanate
Liu KM, et al.
Chemical Communications (Cambridge, England), 51(22), 4655-4658 (2015)
Iron-Catalyzed Room Temperature Cross-Couplings of Bromophenols with Aryl Grignard Reagents
Xu L, et al.
advanced synthesis and catalysis, 361(23), 5421-5427 (2019)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service