Skip to Content
Merck
All Photos(1)

Key Documents

672890

Sigma-Aldrich

3,4-Dichlorophenylacetylene

97%

Synonym(s):

1,2-Dichloro-4-ethynylbenzene, 3,4-Dichloro-1-ethynylbenzene

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$135.00

$135.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 G
$135.00

About This Item

Empirical Formula (Hill Notation):
C8H4Cl2
CAS Number:
Molecular Weight:
171.02
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$135.00


Available to ship on22 April 2025Details


Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

38-42 °C

functional group

chloro

SMILES string

Clc1ccc(cc1Cl)C#C

InChI

1S/C8H4Cl2/c1-2-6-3-4-7(9)8(10)5-6/h1,3-5H

InChI key

MJHFEHXRVNTQIL-UHFFFAOYSA-N

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

205.0 °F - closed cup

Flash Point(C)

96.1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service