Skip to Content
Merck
All Photos(1)

Key Documents

672114

Sigma-Aldrich

Diisopropyl chlorophosphate

97%

Synonym(s):

Diisopropyl phosphorochloridate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$435.00

$435.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$435.00

About This Item

Empirical Formula (Hill Notation):
C6H14ClO3P
CAS Number:
Molecular Weight:
200.60
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$435.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥96.5% (T)
97%

refractive index

n20/D 1.417

SMILES string

CC(C)OP(Cl)(=O)OC(C)C

InChI

1S/C6H14ClO3P/c1-5(2)9-11(7,8)10-6(3)4/h5-6H,1-4H3

InChI key

GUFGWNUUDBGEGH-UHFFFAOYSA-N

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service