Skip to Content
Merck
All Photos(1)

Key Documents

671657

Sigma-Aldrich

1-Ethyl-3-methylimidazolium dimethyl phosphate

≥98.0% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$59.40
50 G
$481.00

$59.40


0 left in stock

Request a Bulk Order

Select a Size

Change View
5 G
$59.40
50 G
$481.00

About This Item

Empirical Formula (Hill Notation):
C8H17N2O4P
CAS Number:
Molecular Weight:
236.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$59.40


0 left in stock

Request a Bulk Order

Quality Level

Assay

≥98.0% (HPLC)

impurities

≤1.0% water

functional group

phosphate

SMILES string

COP([O-])(=O)OC.CCn1cc[n+](C)c1

InChI

1S/C6H11N2.C2H7O4P/c1-3-8-5-4-7(2)6-8;1-5-7(3,4)6-2/h4-6H,3H2,1-2H3;1-2H3,(H,3,4)/q+1;/p-1

InChI key

WTKUDOCGUOSPGV-UHFFFAOYSA-M

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vishal Sharma et al.
Bioresource technology, 285, 121319-121319 (2019-04-14)
Ionic liquid (IL) pretreatment represents an effective strategy for effective fractionation of lignocellulosic biomass (LB) to fermentable sugars in a biorefinery. Optimization of combinatorial pretreatment of sugarcane bagasse (SCB) with IL (1-butyl-3-methylimidazolium chloride [Bmim]Cl) and surfactant (PEG-8000) resulted in enhanced
Parushi Nargotra et al.
Bioresource technology, 289, 121611-121611 (2019-06-18)
The current study presents the first ever report of surfactant (Tween-20) assisted ionic liquid IL, (1-ethyl-3-methylimidazolium methane sulphonate [Emim][MeSO3]) pretreatment of Parthenium hysterophorus biomass, its saccharification by in-house developed enzyme cocktail from Aspergillus aculeatus PN14, and fermentation of sugars to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service