Skip to Content
Merck
All Photos(1)

Key Documents

671134

Sigma-Aldrich

2-Chloro-5-isocyanatopyridine

99%

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$313.00

$313.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$313.00

About This Item

Empirical Formula (Hill Notation):
C6H3ClN2O
CAS Number:
Molecular Weight:
154.55
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$313.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (GC)
98.5-101.5% (argentometric, T)
99%

functional group

chloro
isocyanate

SMILES string

Clc1ccc(cn1)N=C=O

InChI

1S/C6H3ClN2O/c7-6-2-1-5(3-8-6)9-4-10/h1-3H

InChI key

WHENXACISRCOHK-UHFFFAOYSA-N

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service