Skip to Content
Merck
All Photos(1)

Key Documents

669695

Sigma-Aldrich

Ethyl fumaroyl chloride

≥97.0%

Synonym(s):

(E)-3-(Chloroformyl)-acrylic acid ethyl ester, Ethyl fumaryl chloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$161.00
5 G
$641.00

$161.00


Estimated to ship on17 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 G
$161.00
5 G
$641.00

About This Item

Empirical Formula (Hill Notation):
C6H7ClO3
CAS Number:
Molecular Weight:
162.57
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$161.00


Estimated to ship on17 April 2025Details


Request a Bulk Order

Assay

≥97.0%

form

liquid

refractive index

n20/D 1.463

functional group

acyl chloride
ester

SMILES string

CCOC(=O)\C=C\C(Cl)=O

InChI

1S/C6H7ClO3/c1-2-10-6(9)4-3-5(7)8/h3-4H,2H2,1H3/b4-3+

InChI key

YYLWXDIGYFPUSK-ONEGZZNKSA-N

Related Categories

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service