Skip to Content
Merck
All Photos(1)

Key Documents

667315

Sigma-Aldrich

Chloro(η2-P,C-tris(2,4-di-tert-butylphenyl)phosphite)(tricyclohexylphosphine)palladium(II)

Synonym(s):

SamCat, [2-[[Bis[2,4-bis(1,1-dimethylethyl)phenoxy]phosphino-KP]oxy]-3,5-bis(1,1-dimethylethyl)phenyl-KC]chloro(tricyclohexylphosphine)palladium

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$666.00

$666.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$666.00

About This Item

Empirical Formula (Hill Notation):
C60H95ClO3P2Pd
CAS Number:
Molecular Weight:
1068.22
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$666.00


Check Cart for Availability

Request a Bulk Order

form

solid

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

176.7-187.1 °C

SMILES string

C1CCC(CC1)P(C2CCCCC2)C3CCCCC3.CC(C)(C)c4ccc(OP(Oc5ccc(cc5C(C)(C)C)C(C)(C)C)Oc6c([Pd]Cl)cc(cc6C(C)(C)C)C(C)(C)C)c(c4)C(C)(C)C

InChI

1S/C42H62O3P.C18H33P.ClH.Pd/c1-37(2,3)28-19-22-34(31(25-28)40(10,11)12)43-46(44-35-23-20-29(38(4,5)6)26-32(35)41(13,14)15)45-36-24-21-30(39(7,8)9)27-33(36)42(16,17)18;1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;;/h19-23,25-27H,1-18H3;16-18H,1-15H2;1H;/q;;;+1/p-1

InChI key

KSWCAUMOXBNXFL-UHFFFAOYSA-M

Application

Catalyst for coupling of alkyl bromides with aryl boronic acids

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Palladacyclic catalysts by Bedford's group facilitate coupling reactions with low catalyst loadings and challenging aryl chlorides.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service