Skip to Content
Merck
All Photos(1)

Documents

665711

Sigma-Aldrich

4-Pentylphenyl 4-(octyloxy)benzoate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H36O3
CAS Number:
Molecular Weight:
396.56
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

97%

form

liquid crystal

mp

53-57 °C

storage temp.

−20°C

SMILES string

CCCCCCCCOc1ccc(cc1)C(=O)Oc2ccc(CCCCC)cc2

InChI

1S/C26H36O3/c1-3-5-7-8-9-11-21-28-24-19-15-23(16-20-24)26(27)29-25-17-13-22(14-18-25)12-10-6-4-2/h13-20H,3-12,21H2,1-2H3

InChI key

BDTRDPSFINNZRW-UHFFFAOYSA-N

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service