663387
2-Amino-5-(4-chlorophenyl)-1,3,4-oxadiazole
95%
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C8H6ClN3O
CAS Number:
Molecular Weight:
195.61
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay
95%
form
solid
mp
245 °C (dec.)
functional group
chloro
SMILES string
Nc1nnc(o1)-c2ccc(Cl)cc2
InChI
1S/C8H6ClN3O/c9-6-3-1-5(2-4-6)7-11-12-8(10)13-7/h1-4H,(H2,10,12)
InChI key
ZMWWZXSPDQIWAZ-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service