Skip to Content
Merck
All Photos(1)

Key Documents

661732

Sigma-Aldrich

1,3,5-Tris(2-(9-ethylcabazyl-3)ethylene)benzene

Synonym(s):

1,3,5-Tris(2-(9-ethylcabazol-3-yl)ethylene)benzene, TECEB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C54H45N3
CAS Number:
Molecular Weight:
735.96
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

fluorescence

λex 337 nm; λem 416 nm in chloroform

Orbital energy

HOMO 5.2 eV 
LUMO 2.8 eV 

storage temp.

2-8°C

SMILES string

CCn1c2ccccc2c3cc(\C=C\c4cc(\C=C\c5ccc6n(CC)c7ccccc7c6c5)cc(\C=C\c8ccc9n(CC)c%10ccccc%10c9c8)c4)ccc13

InChI

1S/C54H45N3/c1-4-55-49-16-10-7-13-43(49)46-34-37(25-28-52(46)55)19-22-40-31-41(23-20-38-26-29-53-47(35-38)44-14-8-11-17-50(44)56(53)5-2)33-42(32-40)24-21-39-27-30-54-48(36-39)45-15-9-12-18-51(45)57(54)6-3/h7-36H,4-6H2,1-3H3/b22-19+,23-20+,24-21+

InChI key

JWTJCIYHZFCEPU-IKVQWSBMSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charge carrier transporting molecular materials and their applications in devices.
Yasuhiko Shirota et al.
Chemical reviews, 107(4), 953-1010 (2007-04-13)

Articles

Since their discovery, organic light emitting devices (OLEDs) have evolved from a scientific curiosity into a technology with applications in flat panel displays and the potential to revolutionize the lighting market. During their relatively short history, the technology has rapidly advanced, and device efficiencies have increased more than 20-fold, approaching the theoretical limit for internal quantum efficiencies.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service