661570
3-(Trifluoromethyl)phenethylamine hydrochloride
97%
Synonym(s):
3-(Trifluoromethyl)benzeneethanamine hydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C9H11ClF3N
CAS Number:
Molecular Weight:
225.64
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Assay
97%
form
solid
mp
106-110 °C
SMILES string
Cl.NCCc1cccc(c1)C(F)(F)F
InChI
1S/C9H10F3N.ClH/c10-9(11,12)8-3-1-2-7(6-8)4-5-13;/h1-3,6H,4-5,13H2;1H
InChI key
KAFOPDIMPZSSTF-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service