Skip to Content
Merck
All Photos(1)

Key Documents

661082

Sigma-Aldrich

2,4-Difluoropyrimidine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H2F2N2
CAS Number:
Molecular Weight:
116.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.432

bp

118-120 °C

density

1.359 g/mL at 25 °C

functional group

fluoro

SMILES string

Fc1ccnc(F)n1

InChI

1S/C4H2F2N2/c5-3-1-2-7-4(6)8-3/h1-2H

InChI key

JZSYSXZDIQUOGP-UHFFFAOYSA-N

Application

2,4-Difluoropyrimidine can be used as a starting material to synthesize:
  • 3-Pyrimidin-4-yl-oxazolidin-2-one derivatives as mutant isocitrate dehydrogenase (IDH1) inhibitors.[1]
  • N,N′-(1,4-phenylenebis(methylene))bis(2-fluoropyrimidin-4-amine) as chemokine receptor type 4 antagonists.[2]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

106.0 °F - closed cup

Flash Point(C)

41.1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dipyrimidine amines: a novel class of chemokine receptor type 4 antagonists with high specificity
Zhu A, et al.
Journal of Medicinal Chemistry, 53(24), 8556-8568 (2010)
Optimization of 3-pyrimidin-4-yl-oxazolidin-2-ones as orally bioavailable and brain penetrant mutant IDH1 inhibitors
Zhao Q, et al.
ACS Medicinal Chemistry Letters, 9(7), 746-751 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service