Skip to Content
Merck
All Photos(2)

Key Documents

660140

Sigma-Aldrich

NaK Silica gel (K2Na) Stage I

extent of labeling: ~35 wt. % Na-K loading

Synonym(s):

K2Na-SG-I, Sodium-potassium alloy on silica gel (K2Na)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
11101527
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

reagent type: reductant

extent of labeling

~35 wt. % Na-K loading

InChI

1S/K.Na

InChI key

BITYAPCSNKJESK-UHFFFAOYSA-N

Application

Alternative to other reagents for desulfurizations, dehalogenations(as Wurtz coupling reactions), and Birch reductions.[1]

Additionally can be used as:
  • A reducing agent for aliphatic ester substrates[2]
  • A deprotectant in place of the typical heavy metal deprotectant[3]

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - Water-react. 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials, which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P. Nandi, et al.,
Tetrahedron Letters, 50, 3864-3866 (2009)
Dye, J. L. et al
Journal of the American Chemical Society, 127, 9339-9339 (2005)
P. F. Vogt and B. Bodnar,
Spec. Chem., 29, 22-24 (2009)

Articles

SiGNa Chemistry has recently developed a technology for encapsulating alkali metals into nano-structured porous oxides to create stable, free flowing powders with varying reactivities (based on the intended application).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service