Skip to Content
Merck
All Photos(1)

Key Documents

659320

Sigma-Aldrich

3-Iodo-4-nitrotoluene

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$155.00
25 G
$576.00

$155.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
$155.00
25 G
$576.00

About This Item

Linear Formula:
O2NC6H3(I)CH3
CAS Number:
Molecular Weight:
263.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$155.00


Estimated to ship on21 May 2025


Request a Bulk Order

Assay

97%

refractive index

n20/D 1.637

bp

295 °C

density

1.862 g/mL at 25 °C

functional group

iodo
nitro

SMILES string

Cc1ccc(c(I)c1)[N+]([O-])=O

InChI

1S/C7H6INO2/c1-5-2-3-7(9(10)11)6(8)4-5/h2-4H,1H3

InChI key

DJDSQNLIZRLTHJ-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service