Skip to Content
Merck
All Photos(1)

Key Documents

658847

Sigma-Aldrich

(1,2-Methanofullerene C60)-61-carboxylic acid

97% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$873.00

$873.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$873.00

About This Item

Empirical Formula (Hill Notation):
C62H2O2
CAS Number:
Molecular Weight:
778.68
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

$873.00


Check Cart for Availability

Request a Bulk Order

description

functionalized fullerene

Assay

97% (HPLC)

form

powder

functional group

carboxylic acid

Looking for similar products? Visit Product Comparison Guide

General description

Behavior of the fullerene derivative in non aqueous capillary electrophoresis has been studied.[1]

Application

Reagent for preparation of fullerene derivatives.[2][3] Interaction of C60 fullerence derivative with L-histidine was studied in detail. The quantum chemical calculations were focused on the enantioselective binding between the two.[4] (1,2-Methanofullerene C60)-61-carboxylic acid may find potential application in biomedical and cosmetics.[1][2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F. Diederich et al.
Chemical Society Reviews, 23, 243-243 (1994)
Non-aqueous capillary electrophoresis separation of fullerenes and C60 fullerene derivatives."
Astefanei A, et al.
Analytical and Bioanalytical Chemistry, 404(2), 307-313 (2012)
Computational study of enantioselective interaction between C60 fullerene and its derivatives with L-histidine.
Lala B, et al.
Journal of Molecular Modeling, 13(4), 531-536 (2007)
L. Isaacs et al.
Helvetica Chimica Acta, 76, 2454-2454 (1993)

Articles

Find various photovoltaic and bioscience-based applications of fullerenes.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service