Skip to Content
Merck
All Photos(2)

Key Documents

657646

Sigma-Aldrich

QuadraPure® AEA

100-400 μm particle size, extent of labeling: 1.3 mmol/g loading, 1 % cross-linked with divinylbenzene

Synonym(s):

QuadraPure® Aminoethyl aminobut-2-enoate ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$147.00
25 G
$613.00
100 G
$2,090.00

$147.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$147.00
25 G
$613.00
100 G
$2,090.00

About This Item

MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$147.00


Check Cart for Availability

Request a Bulk Order

Quality Level

crosslinking

1 % cross-linked with divinylbenzene

reaction suitability

reagent type: catalyst
reagent type: chelator

extent of labeling

1.3 mmol/g loading

particle size

100-400 μm

Application

Metal Scavenger: Pd, Rh, V, Cu, Fe,

Legal Information

QuadraPure is a registered trademark of Johnson Matthey Finland Oy

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

QuadraPure Cartridges for Removal of Trace Metal from Reaction Mixtures in Flow
A. Hinchcliffe, et al.
Organic Process Research & Development, 11, 477-481 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service