Skip to Content
Merck
All Photos(1)

Key Documents

657190

Sigma-Aldrich

Poly(sebacic acid), diacetoxy terminated

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$468.00

$468.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$468.00

About This Item

Linear Formula:
C2H3O2 (C10H16O3)n C2H3O
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$468.00


Check Cart for Availability

Request a Bulk Order

form

solid

Quality Level

Application

Biodegradable polymer for drug delivery applications.[1]
Poly(sebacic acid) (diacetoxy-terminated) (PSA) may be used as a coating, to ensure erosion and subsequent drug release on only one direction of the coated biomedical device for the treatment of periodontitis.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design of a multiple drug delivery system directed at periodontitis.
Sundararaj SC, et al.
Biomaterials, 34(34), 8835-8842 (2013)
The degradation rate of polyanhydride (poly (sebacic acid), diacetoxy terminated, PSADT).
Peng Y, et al.
Journal of Wuhan University of Technology-Materials Science Edition, 28(4), 793-797 (2013)
Matt J Kipper et al.
Biomaterials, 23(22), 4405-4412 (2002-09-11)
The fabrication, morphological characterization, and drug release kinetics from microspheres of three bioerodible polyanhydrides, poly[1,6-bis(p-carboxyphenoxy)hexane] (poly(CPH)), poly(sebacic anhydride) (poly(SA)), and the copolymer poly(CPH-co-SA) 50:50 (CPH:SA 50:50) is reported. The fabrication technique yields microspheres with different morphologies for each of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service