Skip to Content
Merck
All Photos(1)

Documents

656992

Sigma-Aldrich

Potassium 2,4-difluorophenyltrifluoroborate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3BF5K
CAS Number:
Molecular Weight:
219.99
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

95%

form

solid

mp

>300 °C (lit.)

SMILES string

[K+].Fc1ccc(c(F)c1)[B-](F)(F)F

InChI

1S/C6H3BF5.K/c8-4-1-2-5(6(9)3-4)7(10,11)12;/h1-3H;/q-1;+1

InChI key

HZKKNPYRMYVEOW-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Potassium trifluoroborates are moisture- and air-stable organoboron reagents suitable for oxidative conditions.

Related Content

The central theme of the Molander group's research is the development of new synthetic methods and their application to the synthesis of organic molecules.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service