Skip to Content
Merck
All Photos(1)

Key Documents

656909

Sigma-Aldrich

trans-2-(3-Methoxyphenyl)vinylboronic acid pinacol ester

96%

Synonym(s):

E-2-(3-Methoxyphenyl)vinylboronic acid pinacol ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21BO3
CAS Number:
Molecular Weight:
260.14
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

mp

36-42 °C (lit.)

SMILES string

COc1cccc(\C=C\B2OC(C)(C)C(C)(C)O2)c1

InChI

1S/C15H21BO3/c1-14(2)15(3,4)19-16(18-14)10-9-12-7-6-8-13(11-12)17-5/h6-11H,1-5H3/b10-9+

InChI key

MYKJZXRWDIWMDR-MDZDMXLPSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service