Skip to Content
Merck
All Photos(1)

Key Documents

655600

Sigma-Aldrich

2-(Aminomethyl)-1-ethylpyrrolidine

97%

Synonym(s):

(1-Ethyl-2-pyrrolidinyl)methylamine, (±)-2-(Aminomethyl)-1-ethylpyrrolidine, 1-(1-Ethylpyrrolidin-2-yl)methanamine, 1-Ethyl-2-(aminomethyl)pyrrolidine, 1-Ethyl-2-pyrrolidinemethylamine, N-[(1-Ethylpyrrolidin-2-yl)methyl]amine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$144.00
25 G
$464.00

$144.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$144.00
25 G
$464.00

About This Item

Empirical Formula (Hill Notation):
C7H16N2
CAS Number:
Molecular Weight:
128.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$144.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

refractive index

n20/D 1.466 (lit.)

bp

58-60 °C/16 mmHg (lit.)

density

0.884 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CCN1CCCC1CN

InChI

1S/C7H16N2/c1-2-9-5-3-4-7(9)6-8/h7H,2-6,8H2,1H3

InChI key

UNRBEYYLYRXYCG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

132.8 °F - closed cup

Flash Point(C)

56 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service