Skip to Content
Merck
All Photos(2)

Key Documents

653586

Sigma-Aldrich

4-Fluoro-1,2-phenylenediamine

97%

Synonym(s):

1,2-Diamino-4-fluorobenzene, 4-Fluoro-o-phenylenediamine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$189.00

$189.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$189.00

About This Item

Empirical Formula (Hill Notation):
C6H7FN2
CAS Number:
Molecular Weight:
126.13
Beilstein:
2081075
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$189.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

mp

94-98 °C

functional group

fluoro

SMILES string

Nc1ccc(F)cc1N

InChI

1S/C6H7FN2/c7-4-1-2-5(8)6(9)3-4/h1-3H,8-9H2

InChI key

KWEWNOOZQVJONF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anshul Singh et al.
Bioinorganic chemistry and applications, 2018, 2467463-2467463 (2018-06-01)
We present an inclusive characterization of the unimetallic and heterobimetallic complexes of copper synthesized using CuCl2 and diamine (4-fluoro 1,2-phenylenediamine) resulting in monometallic complex which further undergoes treatment with organometallic dichlorides of group 4 and 14 in 1 : 2 molar ratio
Andreas Kiontke et al.
Analytical and bioanalytical chemistry, 410(16), 3715-3722 (2018-04-14)
Ambient ionization mass spectrometry (AI-MS), the ionization of samples under ambient conditions, enables fast and simple analysis of samples without or with little sample preparation. Due to their simple construction and low resource consumption, plasma-based ionization methods in particular are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service