Skip to Content
Merck
All Photos(1)

Key Documents

652423

Sigma-Aldrich

Dysprosium(II) iodide

anhydrous, powder, ≥99.9% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
DyI2
CAS Number:
Molecular Weight:
416.31
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

grade

anhydrous

Quality Level

Assay

≥99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: dysprosium

impurities

≤1000.0 ppm Trace Rare Earth Analysis

SMILES string

I[Dy]I

InChI

1S/Dy.2HI/h;2*1H/q+2;;/p-2

InChI key

SFCVMDRIXZSBIC-UHFFFAOYSA-L

Application

Used to generate trichlorosilyl radicals from SiCl4 promoting the cyclotrimerization of alkynes to substituted benzenes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhenyu Zhu et al.
Chemical communications (Cambridge, England), (19), 2066-2068 (2006-06-13)
An efficient method for the formation of SiCl3 radicals by the reaction of abundant and cheaper chlorosilanes with DyI2 has been established, not only demonstrating new distinctive reactivities of solvated DyI2 but also suggesting that the presence of lanthanide ions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service