Skip to Content
Merck
All Photos(1)

Key Documents

651761

Sigma-Aldrich

Negative resist remover I

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$45.80

$45.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
$45.80

About This Item

UNSPSC Code:
12352300
NACRES:
NA.23

$45.80


Check Cart for Availability

Request a Bulk Order

shelf life

1 yr (cool area away from direct sunlight)

Quality Level

pH

<2 (20 °C)

bp

204-304 °C (lit.)

density

1.00 g/mL at 25 °C (lit.)

storage temp.

2-8°C

General description

Available as part of Negative Photoresist kit 654892
Low pH formulation for removal of photoresist cured onto various substrates. Non-corrosive to metals and oxides. Does not contain chlorinated hydrocarbons, chromates, phenol, or phosphates. Performs well at room and elevated temperatures.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 2 - Eye Dam. 1 - Skin Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

204.8 °F

Flash Point(C)

96 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Photoresist kit offers pre-weighed chemical components for lithographic processes, with separate etchants for various substrate choices.

Questions

1–2 of 2 Questions  
  1. For the 65,176-1 Negative Resist Remover, do we also buy H2SO4, or is that already in the 65, 176-1?

    1 answer
    1. This product does not include H2SO4.

      Helpful?

  2. Is this remover available with AZ 5214E which is an IR photo-resist (positive to negative) ?

    1 answer
    1. The photoresist remover AZ 5214E (or others options for positive and negative removal) is not available. Unfortunately, the 651761, which is also included in the Negative Photoresist kit (654892), are the only reagents offered at this time.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service