Skip to Content
Merck
All Photos(1)

Key Documents

648450

Sigma-Aldrich

Methyl 5-(3-aminophenyl)furan-2-carboxylate

97%

Synonym(s):

5-(3-Aminophenyl)furan-2-carboxylic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11NO3
CAS Number:
Molecular Weight:
217.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

102-106 °C (lit.)

SMILES string

COC(=O)c1ccc(o1)-c2cccc(N)c2

InChI

1S/C12H11NO3/c1-15-12(14)11-6-5-10(16-11)8-3-2-4-9(13)7-8/h2-7H,13H2,1H3

InChI key

IZIZPLREWPTKFV-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroshi Suzuki et al.
Scientific reports, 10(1), 16892-16892 (2020-10-11)
Recurrent laryngeal nerve (RLN) injury, in which hoarseness and dysphagia arise as a result of impaired vocal fold movement, is a serious complication. Misdirected regeneration is an issue for functional regeneration. In this study, we demonstrated the effect of TrkA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service