Skip to Content
Merck
All Photos(3)

Key Documents

648434

Sigma-Aldrich

1-Methyl-3-phenylpiperazine

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$69.90
5 G
$259.00

$69.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$69.90
5 G
$259.00

About This Item

Empirical Formula (Hill Notation):
C11H16N2
CAS Number:
Molecular Weight:
176.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$69.90


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

56-60 °C (lit.)

functional group

phenyl

SMILES string

CN1CCNC(C1)c2ccccc2

InChI

1S/C11H16N2/c1-13-8-7-12-11(9-13)10-5-3-2-4-6-10/h2-6,11-12H,7-9H2,1H3

InChI key

IRMBVBDXXYXPEW-UHFFFAOYSA-N

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C A Mini et al.
Toxicology in vitro : an international journal published in association with BIBRA, 71, 105069-105069 (2020-12-15)
Three-dimensional skin models, also named 3D skin models, human skin equivalents (HSEs), or Human Epidermal Equivalents (HEEs), have been increasingly used for chemical assessments in terms of efficacy and safety. Considering this, we developed an HEE model using immortalized HaCaT

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service