648418
(−)-DIP-Chloride™ solution
50-65 wt. % in heptane
Synonym(s):
(−)-B-Chlorodiisopinocampheylborane, (−)-Chlorodiisopinocampheylborane, Chlorobis[(1R,2S,3R,5R)-2,6,6-trimethylbicyclo[3.1.1]hept-3-yl]borane, Diisopinocampheylchloroborane
Select a Size
About This Item
Recommended Products
concentration
50-65 wt. % in heptane
density
0.895 g/mL at 25 °C
SMILES string
C[C@H]1[C@@H](C[C@@H]2C[C@H]1C2(C)C)B(Cl)[C@@H]3C[C@@H]4C[C@H]([C@H]3C)C4(C)C
InChI
1S/C20H34BCl/c1-11-15-7-13(19(15,3)4)9-17(11)21(22)18-10-14-8-16(12(18)2)20(14,5)6/h11-18H,7-10H2,1-6H3/t11-,12-,13+,14+,15-,16-,17-,18-/m1/s1
InChI key
PSEHHVRCDVOTID-VMAIWCPRSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
- Enantioselective reduction of α, β-unsaturated trifluoromethyl ketones into corresponding secondary allylic alcohols.
- Asymmetric reduction of aromatic and aliphatic α-halomethyl ketones to haloalcohols.
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
<-22.0 °F - closed cup
Flash Point(C)
< -30 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service