Skip to Content
Merck
All Photos(2)

Key Documents

646679

Sigma-Aldrich

1,1,2-Trimethyl-3-(4-sulfobutyl)benz[e]indolium, inner salt

Dye content 90 %

Synonym(s):

1,1,2-Trimethyl-3-(4-sulfobutyl)-1H-benz[e]indolium hydroxide, inner salt, 1-(4-Sulfobutyl)-2,3,3-trimethyl-4,5-benzindolium, inner salt

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$336.00

$336.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$336.00

About This Item

Empirical Formula (Hill Notation):
C19H23NO3S
CAS Number:
Molecular Weight:
345.46
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$336.00


Check Cart for Availability

Request a Bulk Order

Assay

>90.0% (HPLC)

form

powder or crystals

composition

Dye content, 90%

mp

245-258 °C

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CC1=[N+](CCCCS([O-])(=O)=O)c2ccc3ccccc3c2C1(C)C

InChI

1S/C19H23NO3S/c1-14-19(2,3)18-16-9-5-4-8-15(16)10-11-17(18)20(14)12-6-7-13-24(21,22)23/h4-5,8-11H,6-7,12-13H2,1-3H3

InChI key

GJCURKVOCXJAIV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service