Skip to Content
Merck
All Photos(1)

Key Documents

645346

Sigma-Aldrich

3-Bromo-5-fluorophenylboronic acid

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5BBrFO2
CAS Number:
Molecular Weight:
218.82
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

≥95%

form

solid

SMILES string

OB(O)c1cc(F)cc(Br)c1

InChI

1S/C6H5BBrFO2/c8-5-1-4(7(10)11)2-6(9)3-5/h1-3,10-11H

InChI key

UVKKALLRVWTDOY-UHFFFAOYSA-N

Application

Reactant for:
  • Preparation of functionalized dihalophenylboronic acid via lithiation of dihalophenyl dioxazaborocines and reaction with electrophiles

Other Notes

Contains varying amounts of anhydride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service