Skip to Content
Merck
All Photos(1)

Documents

644501

Sigma-Aldrich

Dichloromethane

purification grade

Synonym(s):

Methylene chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH2Cl2
CAS Number:
Molecular Weight:
84.93
Beilstein:
1730800
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

grade

purification grade

vapor density

2.9 (vs air)

vapor pressure

24.45 psi ( 55 °C)
6.83 psi ( 20 °C)

Assay

≥99.75%

form

liquid

autoignition temp.

1223 °F

expl. lim.

22 %

evapn. residue

≤0.0005%

refractive index

n20/D 1.424 (lit.)

bp

39.8-40 °C (lit.)

mp

−95 °C (lit.)

density

1.325 g/mL at 25 °C (lit.)

SMILES string

ClCCl

InChI

1S/CH2Cl2/c2-1-3/h1H2

InChI key

YMWUJEATGCHHMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Vitamin B12-mediated hydrodechlorination of dichloromethane by bimetallic Cu/Al particles.
Huang CC, et al.
Chemical Engineering Journal, 273, 413-420 (2015)
Dichloromethane Detection Based on Near-Infrared Absorptive Sensing.
Han JH and Yoon SM
IEEE Journal of Selected Topics in Quantum Electronics, 18(5), 1547-1552 (2012)
Dichloromethane photodegradation using titanium catalysts.
Tanguay JF, et al.
Tanguay JF, et al.
J. Catal., 117(2), 335-347 (1989)
Vibrational Relaxation in Liquid Dichloromethane.
Hunter JL and Dardy HD.
J. Chem. Phys. , 42(8), 2961-2966 (1965)
Broadband dielectric response of dichloromethane.
Hunger J, et al.
Chemical Physics Letters, 471(1), 85-91 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service