Skip to Content
Merck
All Photos(1)

Key Documents

643270

Sigma-Aldrich

Gold coated mica

layer thickness 2000 Å, 99.999% (Au)

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
$1,210.00

$1,210.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 SET
$1,210.00

About This Item

Empirical Formula (Hill Notation):
Au
Molecular Weight:
196.97
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$1,210.00


Check Cart for Availability

Request a Bulk Order

Assay

99.999% (Au)

L × W × thickness

1 in. × 1.5 in. × 150 μm

layer thickness

2000 Å

SMILES string

[Au]

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

The gold surface is highly polycrystalline but shows a tendency towards <111> orientation which can be greatly enhanced by annealing.

Packaging

1 ea = 1 in. × 1.5 in. sheet

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Hybrid organic-inorganic sol-gel materials containing silica were first called “ORMOSILs” in 1984.

Questions

  1. Hello. I would be interested in buying mica substrates coated with gold for growing 2D materials by CVD. I understand that the thickness of the gold film is about 200 nm. What is the roughness of the surface? Does it exhibit large flat terraces? Thank you

    1 answer
    1. The surface roughness value for the gold-coated mica product is 2-4 nm. Flat terrace characteristics have not been evaluated.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service