641804
Perylene-66
Dye content 40 %
Synonym(s):
Dibenzthiopheno-perylene -N,N′-dicyclohexylimide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C48H34N2O4S2
Molecular Weight:
766.92
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
Recommended Products
composition
Dye content, 40%
λmax
549 nm
SMILES string
O=C1N(C2CCCCC2)C(=O)c3c4sc5ccccc5c4c6c7ccc8C(=O)N(C9CCCCC9)C(=O)c%10c8c7c(c%11ccc1c3c6%11)c%12sc%13ccccc%13c%10%12
General description
Photoluminescent dye.
Application
An n-channel organic semiconductor.
Coloring of polymeric materials, daylight fluorescent paints, radiation converters.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Newman, C.R., et al.
Chemistry of Materials, 16, 4436-4436 (2004)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service