Skip to Content
Merck
All Photos(1)

Key Documents

64145

Sigma-Aldrich

3-(Dimethylchlorosilyl)propyl methacrylate

technical, ≥85% (GC), (contains BHT as stabilizer)

Synonym(s):

(3-Methacryloyloxypropyl)dimethyl-chlorosilane

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$596.00

$596.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
$596.00

About This Item

Empirical Formula (Hill Notation):
C9H17ClO2Si
CAS Number:
Molecular Weight:
220.77
Beilstein:
2244691
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$596.00


Check Cart for Availability

Request a Bulk Order

grade

technical

Assay

≥85% (GC)

form

liquid

refractive index

n20/D 1.451

density

1.012 g/mL at 20 °C (lit.)

functional group

ester
methacrylate

SMILES string

CC(=C)C(=O)OCCC[Si](C)(C)Cl

InChI

1S/C9H17ClO2Si/c1-8(2)9(11)12-6-5-7-13(3,4)10/h1,5-7H2,2-4H3

InChI key

OKQXCDUCLYWRHA-UHFFFAOYSA-N

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service