Skip to Content
Merck
All Photos(1)

Key Documents

639281

Sigma-Aldrich

Indium tin oxide coated PET

surface resistivity 100 Ω/sq, L × W × thickness 1 ft × 1 ft × 5 mil, sheet

Synonym(s):

ITO-PET, Polyethylene terephthalate film, ITO coated

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
In2O3 · (SnO2)x
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

sheet

Quality Level

surface resistivity

100 Ω/sq

L × W × thickness

1 ft × 1 ft × 5 mil

transmittance

550 nm, >77.0%

SMILES string

O=[In]O[In]=O.O=[Sn]=O

InChI

1S/2In.5O.Sn

InChI key

LNNWKAUHKIHCKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Indium tin oxide coated polyethylene terephthalate (PET) film is optically transparent and electrically conductive.The ITO coating is protected by a thin, transparent film, that can be easily peeled off.

Application

A radius of curvature of up to 75 mm will not damage the ITO coating. Carbon nanomaterial coated ITO –PET offer a new class of transparent organic light emitting diodes. ITO-PET was used as a magnetic target for MALDI.

Features and Benefits

The polyester film products comprise sputtered ITO, and exhibit transmittance and resistance characteristics similar to those of the glass substrates.

Physical properties

Thickness of ITO coating is 720 Å

Quantity

5 ea = 5 x 1ft x 1ft

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A new parameter based on graphene for characterizing transparent, conductive materials
Eigler S, et al.
Carbon, 47(12), 2936-2939 (2009)
Transparent, conductive, and flexible carbon nanotube films and their application in organic light-emitting diodes.
Zhang D, et al.
Nano Letters, 6(9), 1880-1886 (2006)
Modular mass spectrometric tool for analysis of composition and phosphorylation of protein complexes
Blethrow JD, et al.
PLoS ONE, 2(4), e358-e358 (2007)

Articles

In this article, we demonstrate that bis-styrylbenzene derivatives show promising characteristics for very low lasing thresholds and discuss the design considerations for organic lasing molecules.

Organic photovoltaics (OPVs) represent a low-cost, lightweight, and scalable alternative to conventional solar cells. While significant progress has been made in the development of conventional bulk heterojunction cells, new approaches are required to achieve the performance and stability necessary to enable commercially successful OPVs.

A transparent conductive electrode (TCE) is an essential component of various optoelectronic devices such as solar cells, liquid-crystal displays (LCD), light-emitting diodes (LED), and touch screens.

Progress in solution-processed functional materials leads to thin-film optoelectronic devices for industrial and consumer electronics.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service