Skip to Content
Merck
All Photos(1)

Documents

637300

Sigma-Aldrich

Ytterbium(III) oxide

nanopowder, <100 nm particle size (BET), ≥99.7% trace metals basis

Synonym(s):

Diytterbium trioxide, Ytterbia, Ytterbium sesquioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Yb2O3
CAS Number:
Molecular Weight:
394.08
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.7% trace metals basis

form

nanopowder

reaction suitability

reagent type: catalyst
core: ytterbium

particle size

<100 nm (BET)

density

9.17 g/mL at 25 °C

SMILES string

O=[Yb]O[Yb]=O

InChI

1S/3O.2Yb

InChI key

FIXNOXLJNSSSLJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhengze Yu et al.
ACS nano, 9(11), 11064-11074 (2015-10-13)
Photodynamic therapy (PDT) is a well-established modality for cancer therapy, which locally kills cancer cells when light irradiates a photosensitizer. However, conventional PDT is often limited by the extremely short lifespan and severely limited diffusion distance of reactive oxygen species

Articles

Magnetic materials find diverse applications from data storage to renewable energy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service