Skip to Content
Merck
All Photos(1)

Key Documents

636444

Sigma-Aldrich

Diamond

nanopowder, <10 nm particle size (TEM), ≥95% trace metals basis

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$632.00

$632.00


Available to ship on15 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 G
$632.00

About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
EC Number:
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23

$632.00


Available to ship on15 April 2025Details


Request a Bulk Order

Quality Level

Assay

≥95% trace metals basis

form

nanopowder
spherical

surface area

278-335 m2/g , BET

particle size

<10 nm (TEM)

density

3.5 g/mL at 25 °C (lit.)

bulk density

0.17 g/mL

Looking for similar products? Visit Product Comparison Guide

General description

Diamond nanopowder (DNPs) may be produced by multicathode direct current plasma chemical vapor deposition[1] and high pressure high temperature (HPHT).[2]
Potential uses of nano-diamond are in biosensor applications. Surface modification of nano-diamond may enhance its linking mechanism with specific biomolecules.[2] DNPs can be subjected to functionalization post Fenton treatment. These functionalized DNPs may be used as gene delivery systems.[3]

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nano-jewels in biology. Gold and platinum on diamond nanoparticles as antioxidant systems against cellular oxidative stress.
Marti?n R, et al.
ACS Nano, 4(11), 6957-6965 (2010)
Functionalized diamond nanopowder for phosphopeptides enrichment from complex biological fluids.
Hussain D, et al.
Analytica Chimica Acta, 775, 75-84 (2013)
Reactions of amines with CVD diamond nanopowders.
Lee J K, et al.
Diamond and Related Materials, 14(3-7), 675-678 (2005)
Elżbieta Czarniewska et al.
Scientific reports, 9(1), 10330-10330 (2019-07-19)
This article shows that nanodiamonds can transmigrate through the insect cuticle easily, and the doses used were not hemocytotoxic and did not cause inhibition of cellular and humoral immune responses in larvae, pupae and adults of Tenebrio molitor. The examination
Magnetic field imaging and more.
Daniel Evanko
Nature methods, 10(6), 468-468 (2013-07-20)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service