Skip to Content
Merck
All Photos(1)

Key Documents

636185

Sigma-Aldrich

Ethyl 2,3-epoxypropionate

90%

Synonym(s):

Ethyl 2,3-epoxypropanoate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$281.00

$281.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$281.00

About This Item

Empirical Formula (Hill Notation):
C5H8O3
CAS Number:
Molecular Weight:
116.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$281.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

90%

refractive index

n20/D 1.4200 (lit.)

bp

164-165 °C (lit.)

density

1.094 g/mL at 25 °C (lit.)

functional group

ester
ether

storage temp.

2-8°C

SMILES string

CCOC(=O)C1CO1

InChI

1S/C5H8O3/c1-2-7-5(6)4-3-8-4/h4H,2-3H2,1H3

InChI key

LSGWSXRILNPXKJ-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

118.4 - 120.2 °F

Flash Point(C)

48 - 49 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service