Skip to Content
Merck
All Photos(2)

Key Documents

632589

Sigma-Aldrich

2-(2-Hydroxyphenyl)benzothiazole

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$68.70
25 G
$126.00

$68.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$68.70
25 G
$126.00

About This Item

Empirical Formula (Hill Notation):
C13H9NOS
CAS Number:
Molecular Weight:
227.28
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$68.70


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

128-132 °C (lit.)

SMILES string

Oc1ccccc1-c2nc3ccccc3s2

InChI

1S/C13H9NOS/c15-11-7-3-1-5-9(11)13-14-10-6-2-4-8-12(10)16-13/h1-8,15H

InChI key

MVVGSPCXHRFDDR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi-Ping Tong et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 78(2), 809-817 (2011-01-11)
Two Ga(III) complexes with main ligand, 2-(2-hydroxyphenyl)benzothiazole (HL'), namely mixed-ligand ML2X-type [GaL'2X'] (1) (HX'=acetic acid, as ancillary ligand) and the meridianal tris-chelate [GaL'3] (2) have been investigated by the density functional theory (DFT/TDDFT) level calculations. Both 1 and 2 can
Junghwa Lee et al.
The journal of physical chemistry. A, 117(7), 1400-1405 (2013-02-05)
Proton transfer is one of the most important elementary reactions in chemistry and biology. The role of proton in the course of proton transfer, whether it is active or passive, has been the subject of intense investigations. Here we demonstrate
Nawee Kungwan et al.
Physical chemistry chemical physics : PCCP, 14(25), 9016-9025 (2012-04-13)
The dynamics of the excited-state proton transfer (ESPT) in a cluster of 2-(2'-hydroxyphenyl)benzothiazole (HBT) and hydrogen-bonded water molecules was investigated by means of quantum chemical simulations. Two different enol ground-state structures of HBT interacting with the water cluster were chosen
Jie Ma et al.
Chemical communications (Cambridge, England), 48(78), 9720-9722 (2012-08-24)
A new excited state intramolecular proton transfer chromophore of naphthalimide (NI) conjugated 2-(2-hydroxyphenyl) benzothiazole (HBT) was prepared which shows red shifted absorption and long-lived triplet excited states.
Zheng Xu et al.
Chemical communications (Cambridge, England), 48(88), 10871-10873 (2012-10-02)
A new ratiometric fluorescence probe E1 based on an excited-state intramolecular proton transfer (ESIPT) mechanism for detection of hydrogen sulfide (H(2)S) is reported. E1 responds to H(2)S quickly and showed a 30-fold fluorescence enhancement in 2 minutes. Moreover, E1 can

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service