Skip to Content
Merck
All Photos(3)

Documents

632198

Sigma-Aldrich

4-[N,N-Bis(2-hydroxyethyl)amino]benzaldehyde

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HOCH2CH2)2NC6H4CHO
CAS Number:
Molecular Weight:
209.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

55-62 °C (lit.)

SMILES string

OCCN(CCO)c1ccc(C=O)cc1

InChI

1S/C11H15NO3/c13-7-5-12(6-8-14)11-3-1-10(9-15)2-4-11/h1-4,9,13-14H,5-8H2

InChI key

YSDDPNWGLSGZRC-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Janet Sabina Xavier et al.
Bioorganic chemistry, 102, 104081-104081 (2020-07-28)
A new series of thiosemicarbazones were designed and synthesized. Their structures were confirmed by spectral characterization and single crystal XRD studies. Compounds MTSC-2 and ETSC-3 crystallized in the orthorhombic crystal system with space group Pbc21 andPca21respectively. Density functional theory computational

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service