Skip to Content
Merck
All Photos(1)

Key Documents

63207

Sigma-Aldrich

(S)-1-Boc-3-methylpiperazine

≥98%

Synonym(s):

tert-Butyl (S)-3-methyl-1-piperazinecarboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$54.10
5 G
$161.00

$54.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$54.10
5 G
$161.00

About This Item

Empirical Formula (Hill Notation):
C10H20N2O2
CAS Number:
Molecular Weight:
200.28
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$54.10


Check Cart for Availability

Request a Bulk Order

Assay

≥98%

form

solid

optical activity

[α]/D -16±2°, c = 1 in dioxane

SMILES string

C[C@H]1CN(CCN1)C(=O)OC(C)(C)C

InChI

1S/C10H20N2O2/c1-8-7-12(6-5-11-8)9(13)14-10(2,3)4/h8,11H,5-7H2,1-4H3/t8-/m0/s1

InChI key

FMLPQHJYUZTHQS-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in the synthesis of biologically activy molecules including:
  • CCR5 antagonists with anti-HIV-1 activity
  • Opioid receptor antagonists
  • Human growth hormone secretagogue receptor antagonists for treatment of obesity
  • Fatty acid oxidation inhibitors

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service