Skip to Content
Merck
All Photos(1)

Documents

62562

Sigma-Aldrich

Lithium methoxide solution

~10% in methanol (T, ~2.2 M)

Synonym(s):

Lithium methylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OLi
CAS Number:
Molecular Weight:
37.97
Beilstein:
3552389
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

concentration

~10% in methanol (T, ~2.2 M)

density

0.85 g/mL at 20 °C

SMILES string

[Li+].C[O-]

InChI

1S/CH3O.Li/c1-2;/h1H3;/q-1;+1

InChI key

JILPJDVXYVTZDQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Lithium methoxide solution is a useful reagent in organic synthesis with a range of applications. For example, it is used as a strong base in deprotonation reactions of various acidic functional groups, such as alcohols, phenols, and carboxylic acids. It also acts as a catalyst in esterification reactions. It can catalyze the reaction of carboxylic acids with alcohols to form esters.

Application

Lithium methoxide solution (10% in methanol) can be used as a source of lithium ions in the synthesis of dilithium (2,3-dilithium-oxy)-terephthalate, which is studied for its electrochemical behavior.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Self-heat. 1 - Skin Corr. 1B - STOT SE 1

Target Organs

Eyes,Central nervous system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 2

Flash Point(F)

53.6 °F

Flash Point(C)

12 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service