Skip to Content
Merck
All Photos(1)

Key Documents

616435

Sigma-Aldrich

5-Ethyl-5-(4-hydroxyphenyl)-3-methyl-d3 hydantoin

99 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12D3H10N2O3
Molecular Weight:
236.26
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99 atom % D

mass shift

M+3

SMILES string

[2H]C([2H])([2H])N1C(=O)NC(CC)(C1=O)c2ccc(O)cc2

InChI

1S/C12H14N2O3/c1-3-12(8-4-6-9(15)7-5-8)10(16)14(2)11(17)13-12/h4-7,15H,3H2,1-2H3,(H,13,17)/i2D3

InChI key

OQPLORUDZLXXPD-BMSJAHLVSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service