Skip to Content
Merck
All Photos(1)

Key Documents

609382

Sigma-Aldrich

Ammonium-15N calcium nitrate-15N3

5 atom % 15N

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
15NH4Ca(15NO3)3
Molecular Weight:
248.10
UNSPSC Code:
12352116
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

description

Molar Mix 1:1

isotopic purity

5 atom % 15N

mol wt

244.32 by atom % calculation (5 atom % 15N)

mass shift

M+4

SMILES string

[15NH3].[Ca++].O[15N+]([O-])=O.[O-][15N+]([O-])=O.[O-][15N+]([O-])=O

InChI

1S/Ca.HNO3.2NO3.H3N/c;3*2-1(3)4;/h;(H,2,3,4);;;1H3/q+2;;2*-1;/i;4*1+1

InChI key

QGPZBKDRJAXSRG-NJJOSUKLSA-N

General description

Ammonium-15N calcium nitrate-15N3, is enriched with 15N isotope.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service