Skip to Content
Merck
All Photos(1)

Key Documents

606332

Sigma-Aldrich

Hexachlorobenzene-13C6

≥99 atom % 13C, 99%

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$992.00

$992.00


Check Cart for Availability

Request more information

Select a Size

Change View
10 MG
$992.00

About This Item

Linear Formula:
13C6Cl6
CAS Number:
Molecular Weight:
290.74
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.12

$992.00


Check Cart for Availability

Request more information

isotopic purity

≥99 atom % 13C

Assay

99%

form

solid

mass shift

M+6

SMILES string

Cl[13c]1[13c](Cl)[13c](Cl)[13c](Cl)[13c](Cl)[13c]1Cl

InChI

1S/C6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9/i1+1,2+1,3+1,4+1,5+1,6+1

InChI key

CKAPSXZOOQJIBF-IDEBNGHGSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Colette S Azandjeme et al.
Environment international, 69, 1-8 (2014-04-29)
The Borgou region of northern Benin is a major cotton producing area and consistently uses higher amounts of pesticides than other areas of the country. Organochlorine pesticides (OCPs), poorly handled, have been widely used and are still illegally present. We

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service