Skip to Content
Merck
All Photos(1)

Key Documents

60604

Sigma-Aldrich

Potassium p-toluenethiosulfonate

≥97.0% (S)

Synonym(s):

p-Toluenethiosulfonic acid potassium salt, Potassium 4-methylbenzenethiosulfonate, Potassium thiotosylate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$416.00

$416.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$416.00

About This Item

Linear Formula:
CH3C6H4SO2SK
CAS Number:
Molecular Weight:
226.36
Beilstein:
3747559
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder

$416.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥97.0% (S)

form

powder

mp

227-229 °C (lit.)

SMILES string

[K+].Cc1ccc(cc1)S([O-])(=O)=S

InChI

1S/C7H8O2S2.K/c1-6-2-4-7(5-3-6)11(8,9)10;/h2-5H,1H3,(H,8,9,10);/q;+1/p-1

InChI key

RUDNWZFWWJFUSF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service