Skip to Content
Merck
All Photos(1)

Key Documents

604925

Sigma-Aldrich

Creatine-(methyl-13C) monohydrate

99 atom % 13C

Synonym(s):

4-Amidinosarcosine-methyl-13C monohydrate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$2,490.00

$2,490.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 G
$2,490.00

About This Item

Empirical Formula (Hill Notation):
13CC3H9O2N3 · H2O
CAS Number:
Molecular Weight:
150.14
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

$2,490.00


Check Cart for Availability

Request more information

isotopic purity

99 atom % 13C

form

solid

technique(s)

mass spectrometry (MS): suitable

mp

292 °C (dec.) (lit.)

mass shift

M+1

SMILES string

[H]O[H].[13CH3]N(CC(O)=O)C(N)=N

InChI

1S/C4H9N3O2.H2O/c1-7(4(5)6)2-3(8)9;/h2H2,1H3,(H3,5,6)(H,8,9);1H2/i1+1;

InChI key

MEJYXFHCRXAUIL-YTBWXGASSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service