Skip to Content
Merck
All Photos(1)

Key Documents

596701

Sigma-Aldrich

1-(4-Chlorobenzoyl)-1H-benzotriazole

97%

Synonym(s):

1H-Benzotriazol-1-yl(4-chlorophenyl)methanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H8ClN3O
CAS Number:
Molecular Weight:
257.68
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reaction type: C-C Bond Formation

mp

135-139 °C (lit.)

SMILES string

Clc1ccc(cc1)C(=O)n2nnc3ccccc23

InChI

1S/C13H8ClN3O/c14-10-7-5-9(6-8-10)13(18)17-12-4-2-1-3-11(12)15-16-17/h1-8H

InChI key

BLJBQVQHDXUDTE-UHFFFAOYSA-N

Application

Reactant for:
  • Acylation of sodium azide with N-acylbenzotiazoles
  • Gas-phase thermolysis
  • Chemoselective reduction

Reactant for synthesis of:
  • Diketone oximes by phenylation
  • Amides by acylation of amines
  • 1,2-Diketones by coupling of N-acylbenzotriazoles

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service