Skip to Content
Merck
All Photos(2)

Key Documents

594458

Sigma-Aldrich

4-Methoxyphenyl isocyanide

97%

Synonym(s):

1-Isocyano-4-methoxybenzene, 4-Isocyanoanisole, 4-Methoxy-1-isocyanobenzene, 4-Methoxyphenyl isonitrile, Anisyl isonitrile, p-Anisyl isocyanide, p-Anisyl isonitrile, p-Methoxyphenyl isocyanide, p-Methoxyphenyl isonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$152.00

$152.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$152.00

About This Item

Linear Formula:
H3COC6H4NC
CAS Number:
Molecular Weight:
133.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$152.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

mp

29-33 °C (lit.)

storage temp.

−20°C

SMILES string

COc1ccc(cc1)[N+]#[C-]

InChI

1S/C8H7NO/c1-9-7-3-5-8(10-2)6-4-7/h3-6H,2H3

InChI key

MIEQKTRWKZYUPY-UHFFFAOYSA-N

Application

4-Methoxyphenyl isocyanide can be used to synthesize N-(isoquinolin-1-yl)-formamides via reaction with 2-alkynylbenzaldoximes in the presence of silver trifluoromethanesulfonate and bismuth triflate.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Multicatalytic Tandem Reactions of 2-Alkynylbenzaldoximes with Isocyanides"
Chen Z, et al.
AZo Journal of Materials Online, 351(16), 2702- 2708 (2009)
Hideaki Komiyama et al.
Nanotechnology, 26(39), 395302-395302 (2015-09-12)
We present an alternative approach to fabricating hexagonally arranged nanodot arrays of various metals by seed-mediated electroless plating with a cylinder-forming block copolymer thin film, PEO-b-PMA(Az), as a scaffold. Metal ions were selectively incorporated into PEO cylinders, followed by their

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service