Skip to Content
Merck
All Photos(3)

Key Documents

594261

Sigma-Aldrich

5-Bromoisoquinoline

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6BrN
CAS Number:
Molecular Weight:
208.05
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

83-87 °C (lit.)

functional group

bromo

SMILES string

Brc1cccc2cnccc12

InChI

1S/C9H6BrN/c10-9-3-1-2-7-6-11-5-4-8(7)9/h1-6H

InChI key

CYJZJGYYTFQQBY-UHFFFAOYSA-N

Application

Starting material employed in palladium-catalyzed aminomethylation[1] and amination[2] reactions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Qilong Shen et al.
Journal of the American Chemical Society, 128(31), 10028-10029 (2006-08-03)
A mild, palladium-catalyzed coupling of aryl halides with ammonia or lithium amide to form primary arylamines as the major product is described. These reactions occurred with excellent selectivity for formation of the primary arylamine over formation of the diarylamine (9.5:1
Gary A Molander et al.
Organic letters, 9(8), 1597-1600 (2007-03-21)
[reaction: see text] The Suzuki-Miyaura cross-coupling reaction of N,N-dialkylaminomethyltrifluoroborates with aryl halides allows the construction of an aminomethyl aryl linkage through a disconnection based on dissonant reactivity patterns. A variety of these aminomethyltrifluoroborate substrates were prepared in good to excellent

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service