Skip to Content
Merck
All Photos(1)

Documents

592668

Sigma-Aldrich

Iodoacetamide-15N

98 atom % 15N

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ICH2CO15NH2
CAS Number:
Molecular Weight:
185.96
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

98 atom % 15N

form

solid

technique(s)

protein expression: suitable

mp

92-95 °C (lit.)

mass shift

M+1

SMILES string

[15NH2]C(=O)CI

InChI

1S/C2H4INO/c3-1-2(4)5/h1H2,(H2,4,5)/i4+1

InChI key

PGLTVOMIXTUURA-AZXPZELESA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service