Skip to Content
Merck
All Photos(2)

Key Documents

591882

Sigma-Aldrich

(3R)-(−)-3-(1-Methyl-1H-indol-3-yl)butyraldehyde

98%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$1,680.00

$1,680.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$1,680.00

About This Item

Empirical Formula (Hill Notation):
C13H15NO
CAS Number:
Molecular Weight:
201.26
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$1,680.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

optical activity

[α]20/D -6, c = 1% in chloroform

optical purity

ee: 99% (HPLC)

mp

55.5-59.5 °C (lit.)

functional group

aldehyde

SMILES string

C[C@H](CC=O)c1cn(C)c2ccccc12

InChI

1S/C13H15NO/c1-10(7-8-15)12-9-14(2)13-6-4-3-5-11(12)13/h3-6,8-10H,7H2,1-2H3/t10-/m1/s1

InChI key

OQWWHYBHQFZHLP-SNVBAGLBSA-N

Application

(3R)-(−)-3-(1-Methyl-1H-indol-3-yl)butyraldehyde can be used as a substrate in the synthesis of 2-alkyl cyclohexanone intermediates, applicable in the preparation of tricyclic steroid precursors.[1]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A three-step route to a tricyclic steroid precursor
Taber DF and Sheth RB
The Journal of Organic Chemistry, 73(20), 8030-8032 (2008)
Can simple enones be useful partners for the catalytic stereoselective alkylation of indoles?
Bandini M, et al.
The Journal of Organic Chemistry, 69(22), 7511-7518 (2004)
Catalytic enantioselective conjugate addition of indoles to simple α, β-unsaturated ketones
Bandini M, et al.
Tetrahedron Letters, 44(31), 5843-5846 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service